Learn More
Meso Scale Discovery U-PLEX NHP MIP-1 ASY
SDP
Catalog No. 5000080711
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse and rat MIP-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a quick streamlined workflow using an automated plate washer This assay requires 50 L or less sample volume and has a lower limit of detection of 19 5 pg/mL for mouse MIP-1alpha and 156 pg/mL for rat MIP-1alpha
Catalog No. 50-000-80711 Supplier Meso Scale Discovery Supplier No. K156UJK2
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse and rat MIP-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a quick streamlined workflow using an automated plate washer This assay requires 50 L or less sample volume and has a lower limit of detection of 19 5 pg/mL for mouse MIP-1alpha and 156 pg/mL for rat MIP-1alpha