Learn More
Meso Scale Discovery U-PLEX NHP MIP-1B ASY
SDP
Catalog No. 5000080645
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1beta Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MIP-1beta in non-human primate (NHP) samples such as serum plasma and culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for MIP-1beta detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay has a typical lower limit of detection of 1 3 pg/mL and an upper limit of 100 000 pg/mL for MIP-1beta
Catalog No. 50-000-80645 Supplier Meso Scale Discovery Supplier No. K156UKK1
The Meso Scale Discovery U-PLEX NHP MIP-1beta Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MIP-1beta in non-human primate (NHP) samples such as serum plasma and culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies for MIP-1beta detection antibodies calibrators controls 10X diluent read buffer and plate seals The assay has a typical lower limit of detection of 1 3 pg/mL and an upper limit of 100 000 pg/mL for MIP-1beta