Learn More
Meso Scale Discovery U-PLEX NHP MIP-1B ASYQP
SDP
Catalog No. 5000079774
Encompass
The U-PLEX NHP MIP-1 Assay (K156UKK) is a specialized immunoassay kit designed for quantitative measurement of Macrophage Inflammatory Protein-1 beta in non-human primate samples This electrochemiluminescent detection assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with reliable tools for cytokine analysis in preclinical studies involving non-human primate models
Catalog No. 50-000-79774 Supplier Meso Scale Discovery Supplier No. K156UKK21
The U-PLEX NHP MIP-1 Assay (K156UKK) is a specialized immunoassay kit designed for quantitative measurement of Macrophage Inflammatory Protein-1 beta in non-human primate samples This electrochemiluminescent detection assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with reliable tools for cytokine analysis in preclinical studies involving non-human primate models