Learn More
Meso Scale Discovery U-PLEX NHP MIP-3 ASY 25 PL
SDP
Catalog No. 5000080761
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary U-PLEX technology is highly sensitive has minimal matrix interference and requires low sample volume The assay can detect MIP-3 concentrations from 0 04 - 500 pg/mL and has high specificity with no cross-reactivity to other cytokines This product provides a rapid reproducible method to quantify MIP-3 levels for life science research applications
Catalog No. 50-000-80761 Supplier Meso Scale Discovery Supplier No. K156UZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary U-PLEX technology is highly sensitive has minimal matrix interference and requires low sample volume The assay can detect MIP-3 concentrations from 0 04 - 500 pg/mL and has high specificity with no cross-reactivity to other cytokines This product provides a rapid reproducible method to quantify MIP-3 levels for life science research applications