Learn More
Meso Scale Discovery U-PLEX NHP MIP-3 ASY
SDP
Catalog No. 5000080122
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This product provides components for one 96-well plate which can assay 40 samples in duplicate
Catalog No. 50-000-80122 Supplier Meso Scale Discovery Supplier No. K156UZK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This product provides components for one 96-well plate which can assay 40 samples in duplicate