Learn More
Meso Scale Discovery U-PLEX NHP MIP-3 ASY
SDP
Catalog No. 5000080759
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for measuring MIP-3 concentrations
Catalog No. 50-000-80759 Supplier Meso Scale Discovery Supplier No. K156UZK1
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for the quantitative measurement of Macrophage Inflammatory Protein 3 alpha (MIP-3 ) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for measuring MIP-3 concentrations