Learn More
Meso Scale Discovery U-PLEX NHP MIP-3 ASY384-well
SDP
Catalog No. 5000081746
Encompass
The Meso Scale Discovery Non-Human Primate MIP-3 alpha Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures Macrophage Inflammatory Protein 3 alpha (MIP-3 alpha) in non-human primate samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent This assay can be used to measure MIP-3 alpha levels in non-human primate samples for immunology and inflammation research
Catalog No. 50-000-81746 Supplier Meso Scale Discovery Supplier No. K256UZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Non-Human Primate MIP-3 alpha Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures Macrophage Inflammatory Protein 3 alpha (MIP-3 alpha) in non-human primate samples such as serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent This assay can be used to measure MIP-3 alpha levels in non-human primate samples for immunology and inflammation research