Learn More
Meso Scale Discovery U-PLEX NHP MIP-3 ASYQP
SDP
Catalog No. 5000079744
Encompass
The U-PLEX NHP MIP-3 Assay (K156UZK) is a specialized immunoassay kit designed for quantitative measurement of Macrophage Inflammatory Protein-3 alpha in non-human primate samples This electrochemiluminescent detection assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for accurate biomarker analysis in preclinical studies involving non-human primate models
Catalog No. 50-000-79744 Supplier Meso Scale Discovery Supplier No. K156UZK21
The U-PLEX NHP MIP-3 Assay (K156UZK) is a specialized immunoassay kit designed for quantitative measurement of Macrophage Inflammatory Protein-3 alpha in non-human primate samples This electrochemiluminescent detection assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for accurate biomarker analysis in preclinical studies involving non-human primate models