Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASY 25 PL
SDP
Catalog No. 5000080295
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for measuring Macrophage Inflammatory Protein 3 beta (MIP-3 ) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve This assay offers sensitive reproducible quantification of MIP-3 with a broad dynamic range and minimal sample volume
Catalog No. 50-000-80295 Supplier Meso Scale Discovery Supplier No. K156VAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for measuring Macrophage Inflammatory Protein 3 beta (MIP-3 ) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve This assay offers sensitive reproducible quantification of MIP-3 with a broad dynamic range and minimal sample volume