Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASY
SDP
Catalog No. 5000079742
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Macrophage Inflammatory Protein 3 beta (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well or 384-well plate format The kit includes all necessary reagents and calibrators needed to perform the assay Results are obtained in as little as 3 5 hours with minimal sample volume requirements This assay provides researchers with a sensitive and reproducible tool for studying MIP-3 levels across experimental conditions
Catalog No. 50-000-79742 Supplier Meso Scale Discovery Supplier No. K156VAK1
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Macrophage Inflammatory Protein 3 beta (MIP-3 ) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well or 384-well plate format The kit includes all necessary reagents and calibrators needed to perform the assay Results are obtained in as little as 3 5 hours with minimal sample volume requirements This assay provides researchers with a sensitive and reproducible tool for studying MIP-3 levels across experimental conditions