Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASY
SDP
Catalog No. 5000079743
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for measuring Macrophage Inflammatory Protein 3 (MIP-3 ) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve It offers high sensitivity a broad dynamic range and requires only 50 L per well The kit is for research use only
Catalog No. 50-000-79743 Supplier Meso Scale Discovery Supplier No. K156VAK2
The Meso Scale Discovery U-PLEX NHP MIP-3 Assay is a multiplex immunoassay kit for measuring Macrophage Inflammatory Protein 3 (MIP-3 ) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve It offers high sensitivity a broad dynamic range and requires only 50 L per well The kit is for research use only