Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASY384-well
SDP
Catalog No. 5000081747
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-3 384-well Assay Kit is a multiplex immunoassay kit for the quantitative measurement of Macrophage Inflammatory Protein 3 (MIP-3 ) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a sensitivity of 0 08 pg/mL and an assay range from 0 08-500 pg/mL The kit can perform up to 380 tests and has a shelf life of 9 months when stored at 2-8 C This product enables high-throughput sensitive and reproducible quantification of the chemokine MIP-3
Catalog No. 50-000-81747 Supplier Meso Scale Discovery Supplier No. K256VAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-3 384-well Assay Kit is a multiplex immunoassay kit for the quantitative measurement of Macrophage Inflammatory Protein 3 (MIP-3 ) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a sensitivity of 0 08 pg/mL and an assay range from 0 08-500 pg/mL The kit can perform up to 380 tests and has a shelf life of 9 months when stored at 2-8 C This product enables high-throughput sensitive and reproducible quantification of the chemokine MIP-3