Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASYQP
SDP
Catalog No. 5000080294
Encompass
MSD s U-PLEX MIP-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative detection of Macrophage Inflammatory Protein-3 in non-human primate samples This research tool utilizes MSD s proprietary MULTI-SPOT technology for sensitive precise biomarker measurement in serum plasma and other biological matrices The assay offers excellent reproducibility and dynamic range for inflammatory research applications in preclinical studies
Catalog No. 50-000-80294 Supplier Meso Scale Discovery Supplier No. K156VAK21
MSD s U-PLEX MIP-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative detection of Macrophage Inflammatory Protein-3 in non-human primate samples This research tool utilizes MSD s proprietary MULTI-SPOT technology for sensitive precise biomarker measurement in serum plasma and other biological matrices The assay offers excellent reproducibility and dynamic range for inflammatory research applications in preclinical studies