Learn More
Meso Scale Discovery U-PLEX NHP MIP-3B ASYQP
SDP
Catalog No. 5000080635
Encompass
MSD s U-PLEX MIP-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative detection of Macrophage Inflammatory Protein-3 in non-human primate samples This research tool utilizes MSD s proprietary MULTI-SPOT technology for sensitive precise biomarker measurement in serum plasma and other biological matrices The assay offers excellent reproducibility and dynamic range for inflammatory research applications supporting drug development and biomedical studies requiring accurate cytokine quantification in primate models
Catalog No. 50-000-80635 Supplier Meso Scale Discovery Supplier No. K156VAK22
MSD s U-PLEX MIP-3 Singleplex Assay is an electrochemiluminescence-based immunoassay designed for quantitative detection of Macrophage Inflammatory Protein-3 in non-human primate samples This research tool utilizes MSD s proprietary MULTI-SPOT technology for sensitive precise biomarker measurement in serum plasma and other biological matrices The assay offers excellent reproducibility and dynamic range for inflammatory research applications supporting drug development and biomedical studies requiring accurate cytokine quantification in primate models