Learn More
Meso Scale Discovery U-PLEX NHP MIP-5 ASY 25 PL
SDP
Catalog No. 5000078494
Encompass
The Meso Scale Discovery U-PLEX NHP MIP 5 Assay is a multiplex immunoassay panel measuring 5 biomarkers in human serum plasma and CSF It uses electrochemiluminescence detection for A 42 t-tau p-tau -synuclein and NFL offering high sensitivity and a broad dynamic range from a small sample This panel aids in assessing Alzheimer s Parkinson s and related disorders The kit includes a 96-well plate detection antibodies calibrators controls 10X read buffer sample diluent and plate seals for one 96-well plate
Catalog No. 50-000-78494 Supplier Meso Scale Discovery Supplier No. K156XSK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP 5 Assay is a multiplex immunoassay panel measuring 5 biomarkers in human serum plasma and CSF It uses electrochemiluminescence detection for A 42 t-tau p-tau -synuclein and NFL offering high sensitivity and a broad dynamic range from a small sample This panel aids in assessing Alzheimer s Parkinson s and related disorders The kit includes a 96-well plate detection antibodies calibrators controls 10X read buffer sample diluent and plate seals for one 96-well plate