Learn More
Meso Scale Discovery U-PLEX NHP MIP-5 ASY
SDP
Supplier:  Meso Scale Discovery K156XSK2
Encompass
The Meso Scale Discovery U-PLEX NHP MIP 5 Assay is a multiplex immunoassay panel that measures 5 neurodegenerative and neuroinflammatory biomarkers in human serum plasma and CSF samples The assay uses electrochemiluminescent detection technology and provides quantitative results for A 1-42 t-tau p-tau -synuclein and YKL-40 This assay allows researchers to profile these key biomarkers related to Alzheimer s disease Parkinson s disease and neuroinflammation in a single small-volume sample The high sensitivity broad dynamic range and rapid turnaround time of this multiplex panel makes it well-suited for clinical research and drug development applications
Catalog No. 50-000-78619