Learn More
Meso Scale Discovery U-PLEX NHP SDF-1 ASY 25 PL
SDP
Catalog No. 5000080046
Encompass
The U-PLEX NHP SDF-1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Stromal Cell-Derived Factor 1 alpha (SDF-1 ) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows for the rapid reproducible quantification of SDF-1 levels from various biological matrices
Catalog No. 50-000-80046 Supplier Meso Scale Discovery Supplier No. K156VBK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP SDF-1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of human Stromal Cell-Derived Factor 1 alpha (SDF-1 ) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows for the rapid reproducible quantification of SDF-1 levels from various biological matrices