Learn More
Meso Scale Discovery U-PLEX NHP SDF-1 ASY384-well
SDP
Catalog No. 5000080219
Encompass
The Meso Scale Discovery U-PLEX SDF-1alpha Singleplex Assay Kit is a highly sensitive immunoassay for the quantitative measurement of SDF-1alpha in non-human primate samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology on Meso Scale Discovery s proprietary multi-array plate platform The assay has high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-80219 Supplier Meso Scale Discovery Supplier No. K256VBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX SDF-1alpha Singleplex Assay Kit is a highly sensitive immunoassay for the quantitative measurement of SDF-1alpha in non-human primate samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology on Meso Scale Discovery s proprietary multi-array plate platform The assay has high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control The assay takes approximately 3 5 hours to complete