Learn More
Meso Scale Discovery U-PLEX NHP TARC ASY
SDP
Catalog No. 5000079893
Encompass
The U-PLEX NHP TARC Assay Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and can detect TARC in the range of 0 3-2500 pg/mL TARC is a chemokine produced by dendritic cells endothelial cells and keratinocytes and is a biomarker for diseases like atopic dermatitis This assay allows for the accurate quantification of TARC levels in biological samples
Catalog No. 50-000-79893 Supplier Meso Scale Discovery Supplier No. K156ULK2
The U-PLEX NHP TARC Assay Kit from Meso Scale Diagnostics is a highly sensitive immunoassay that quantitatively measures Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and can detect TARC in the range of 0 3-2500 pg/mL TARC is a chemokine produced by dendritic cells endothelial cells and keratinocytes and is a biomarker for diseases like atopic dermatitis This assay allows for the accurate quantification of TARC levels in biological samples