Learn More
Meso Scale Discovery U-PLEX NHP TARC ASY
SDP
Catalog No. 5000080237
Encompass
The U-PLEX NHP TARC Assay Kit from Meso Scale Diagnostics is a sensitive immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a capture antibody-coated plate detection antibody calibrator and diluent It offers high sensitivity and a broad dynamic range requiring only 25 L of sample enabling efficient TARC testing from limited quantities
Catalog No. 50-000-80237 Supplier Meso Scale Discovery Supplier No. K156ULK1
The U-PLEX NHP TARC Assay Kit from Meso Scale Diagnostics is a sensitive immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 96-well plate format The kit includes a capture antibody-coated plate detection antibody calibrator and diluent It offers high sensitivity and a broad dynamic range requiring only 25 L of sample enabling efficient TARC testing from limited quantities