Learn More
Meso Scale Discovery U-PLEX NHP TARC ASY384-well
SDP
Catalog No. 5000081239
Encompass
The U-PLEX NHP TARC 384-well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It includes a 384-well plate with a monoclonal antibody for TARC a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and calibrator vial The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL facilitating high-throughput research on TARC in allergic inflammatory diseases
Catalog No. 50-000-81239 Supplier Meso Scale Discovery Supplier No. K256ULK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP TARC 384-well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring Thymus and Activation Regulated Chemokine (TARC/CCL17) in human serum plasma and cell culture supernatants It includes a 384-well plate with a monoclonal antibody for TARC a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and calibrator vial The assay s sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL facilitating high-throughput research on TARC in allergic inflammatory diseases