Learn More
Meso Scale Discovery U-PLEX NHP TGF-B1 ASY 25 PL
SDP
Catalog No. 5000080743
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta1 Assay Kit is a multiplex immunoassay that measures active and total TGF-beta1 levels in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators and reagents required to perform testing Results are obtained in as little as 3 5 hours This assay allows for the quantitative determination of TGF-beta1 levels in NHP samples
Catalog No. 50-000-80743 Supplier Meso Scale Discovery Supplier No. K156XWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta1 Assay Kit is a multiplex immunoassay that measures active and total TGF-beta1 levels in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a plate coated with capture antibodies detection antibodies calibrators and reagents required to perform testing Results are obtained in as little as 3 5 hours This assay allows for the quantitative determination of TGF-beta1 levels in NHP samples