Learn More
Meso Scale Discovery U-PLEX NHP TGF-B1 ASY
SDP
Catalog No. 5000078620
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta 1 Assay Kit is a multiplex immunoassay for measuring active and total TGF-beta 1 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantitation with high specificity and low background The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals for one 96-well plate enabling accurate TGF-beta 1 measurement in research samples
Catalog No. 50-000-78620 Supplier Meso Scale Discovery Supplier No. K156XWK1
The Meso Scale Discovery U-PLEX NHP TGF-beta 1 Assay Kit is a multiplex immunoassay for measuring active and total TGF-beta 1 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantitation with high specificity and low background The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals for one 96-well plate enabling accurate TGF-beta 1 measurement in research samples