Learn More
Meso Scale Discovery U-PLEX NHP TGF-B1 ASY384-well
SDP
Catalog No. 5000079237
Encompass
The Meso Scale Discovery U-PLEX TGF-beta 1 Kit is a singleplex assay for measuring TGF-beta 1 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive accurate quantification with a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 instrument offering a simple rapid method for research applications
Catalog No. 50-000-79237 Supplier Meso Scale Discovery Supplier No. K256XWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX TGF-beta 1 Kit is a singleplex assay for measuring TGF-beta 1 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive accurate quantification with a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 instrument offering a simple rapid method for research applications