Learn More
Meso Scale Discovery U-PLEX NHP TGF-B1 ASYQP
SDP
Catalog No. 5000078460
Encompass
MSD s U-PLEX TGF 1 assay is a singleplex electrochemiluminescence immunoassay designed for non-human primate samples This ultra-sensitive platform enables precise quantification of transforming growth factor beta-1 a critical cytokine involved in immune regulation and tissue remodeling The U-PLEX technology offers superior sensitivity and dynamic range compared to traditional ELISA methods making it ideal for research applications requiring accurate TGF 1 measurement in primate biological samples
Catalog No. 50-000-78460 Supplier Meso Scale Discovery Supplier No. K156XWK21
MSD s U-PLEX TGF 1 assay is a singleplex electrochemiluminescence immunoassay designed for non-human primate samples This ultra-sensitive platform enables precise quantification of transforming growth factor beta-1 a critical cytokine involved in immune regulation and tissue remodeling The U-PLEX technology offers superior sensitivity and dynamic range compared to traditional ELISA methods making it ideal for research applications requiring accurate TGF 1 measurement in primate biological samples