Learn More
Meso Scale Discovery U-PLEX NHP TGF-B2 ASY 25 PL
SDP
Catalog No. 5000078463
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta 2 Assay is a multiplex immunoassay kit for measuring active and total TGF-beta 2 in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate capture and detection antibodies calibrators and reagents for testing 46 samples in duplicate with a 3 5-hour turnaround This assay enables sensitive reproducible quantitation of TGF-beta 2 a key biomarker
Catalog No. 50-000-78463 Supplier Meso Scale Discovery Supplier No. K156XUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta 2 Assay is a multiplex immunoassay kit for measuring active and total TGF-beta 2 in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate capture and detection antibodies calibrators and reagents for testing 46 samples in duplicate with a 3 5-hour turnaround This assay enables sensitive reproducible quantitation of TGF-beta 2 a key biomarker