Learn More
Meso Scale Discovery U-PLEX NHP TGF-B2 ASY
SDP
Catalog No. 5000078489
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta2 Assay is a multiplex immunoassay kit for measuring active and total TGF-beta2 in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents for testing 46 samples in duplicate Turnaround time is 3 5 hours enabling sensitive reproducible quantification of TGF-beta2 levels
Catalog No. 50-000-78489 Supplier Meso Scale Discovery Supplier No. K156XUK1
The Meso Scale Discovery U-PLEX NHP TGF-beta2 Assay is a multiplex immunoassay kit for measuring active and total TGF-beta2 in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents for testing 46 samples in duplicate Turnaround time is 3 5 hours enabling sensitive reproducible quantification of TGF-beta2 levels