Learn More
Meso Scale Discovery U-PLEX NHP TGF-B2 ASYQP
SDP
Catalog No. 5000078622
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta2 in non-human primate (NHP) samples such as serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the reliable quantification of TGF-beta2 an important biomarker implicated in immune regulation and disease progression With minimal hands-on time this kit serves as a powerful tool for preclinical NHP studies of immune function and disease
Catalog No. 50-000-78622 Supplier Meso Scale Discovery Supplier No. K156XUK22
The Meso Scale Discovery U-PLEX NHP TGF-beta2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta2 in non-human primate (NHP) samples such as serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the reliable quantification of TGF-beta2 an important biomarker implicated in immune regulation and disease progression With minimal hands-on time this kit serves as a powerful tool for preclinical NHP studies of immune function and disease