Learn More
Meso Scale Discovery U-PLEX NHP TGF-B3 ASY 25 PL
SDP
Catalog No. 5000078361
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows sensitive reproducible quantification of TGF-beta3 levels from NHP samples
Catalog No. 50-000-78361 Supplier Meso Scale Discovery Supplier No. K156XVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows sensitive reproducible quantification of TGF-beta3 levels from NHP samples