Learn More
Meso Scale Discovery U-PLEX NHP TGF-B3 ASY
SDP
Catalog No. 5000078363
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of active TGF-beta3 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform testing on 50 microplate wells The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78363 Supplier Meso Scale Discovery Supplier No. K156XVK2
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of active TGF-beta3 in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform testing on 50 microplate wells The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL