Learn More
Meso Scale Discovery U-PLEX NHP TGF-B3 ASYQP
SDP
Catalog No. 5000080781
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible quantification of TGF-beta3 levels
Catalog No. 50-000-80781 Supplier Meso Scale Discovery Supplier No. K156XVK21
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta3 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible quantification of TGF-beta3 levels