Learn More
Meso Scale Discovery U-PLEX NHP TNF- ASY 25 PL
SDP
Catalog No. 5000079904
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-alpha Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TNF-alpha in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity and no cross-reactivity It requires 50 L sample volume and the total assay time is approximately 3 5 hours The lower limit of detection is 0 04 pg/mL This assay provides a rapid reproducible method to measure TNF-alpha for life science research applications
Catalog No. 50-000-79904 Supplier Meso Scale Discovery Supplier No. K156UCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TNF-alpha Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TNF-alpha in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity and no cross-reactivity It requires 50 L sample volume and the total assay time is approximately 3 5 hours The lower limit of detection is 0 04 pg/mL This assay provides a rapid reproducible method to measure TNF-alpha for life science research applications