Learn More
Meso Scale Discovery U-PLEX NHP TNF-B ASY
SDP
Catalog No. 5000080080
Encompass
The Meso Scale Discovery U-PLEX NHP TNF beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of TNF beta in non-human primate serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for NHP TNF beta with no cross-reactivity to other cytokines It includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator The total assay time is approximately 3 5 hours
Catalog No. 50-000-80080 Supplier Meso Scale Discovery Supplier No. K156UUK1
The Meso Scale Discovery U-PLEX NHP TNF beta Assay is an electrochemiluminescent immunoassay for the quantitative measurement of TNF beta in non-human primate serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for NHP TNF beta with no cross-reactivity to other cytokines It includes a plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator The total assay time is approximately 3 5 hours