Learn More
Meso Scale Discovery U-PLEX NHP TNF-B ASY
SDP
Catalog No. 5000080081
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-beta Assay is a multiplex immunoassay kit for quantifying TNF-beta in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology and includes plates with capture antibodies detection antibodies calibrators and diluents The assay is sensitive has a broad dynamic range requires only 25 L of sample and delivers results in 3 5 hours with minimal hands-on time enabling efficient and accurate measurement of NHP TNF-beta levels
Catalog No. 50-000-80081 Supplier Meso Scale Discovery Supplier No. K156UUK2
The Meso Scale Discovery U-PLEX NHP TNF-beta Assay is a multiplex immunoassay kit for quantifying TNF-beta in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology and includes plates with capture antibodies detection antibodies calibrators and diluents The assay is sensitive has a broad dynamic range requires only 25 L of sample and delivers results in 3 5 hours with minimal hands-on time enabling efficient and accurate measurement of NHP TNF-beta levels