Learn More
Meso Scale Discovery U-PLEX NHP TNF-B ASY384-well
SDP
Catalog No. 5000079202
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-beta 384-well Assay Kit is a multiplex immunoassay that measures human TNF-beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows high throughput screening and includes standards controls calibrators detection antibody read buffer and 384-well plates It can detect TNF-beta concentrations as low as 0 04 pg/mL with an upper limit of 10 000 pg/mL The assay takes approximately 3 hours to complete
Catalog No. 50-000-79202 Supplier Meso Scale Discovery Supplier No. K256UUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TNF-beta 384-well Assay Kit is a multiplex immunoassay that measures human TNF-beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This 384-well plate assay allows high throughput screening and includes standards controls calibrators detection antibody read buffer and 384-well plates It can detect TNF-beta concentrations as low as 0 04 pg/mL with an upper limit of 10 000 pg/mL The assay takes approximately 3 hours to complete