Learn More
Meso Scale Discovery U-PLEX NHP TPO ASY 25 PL
SDP
Catalog No. 5000079780
Encompass
The Meso Scale Discovery U-PLEX NHP TPO Assay is an electrochemiluminescent immunoassay for measuring thyroid peroxidase autoantibodies in human serum plasma and whole blood It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The assay has a dynamic range of 0 1-1000 U/mL and requires 25 L per well The kit includes a 96-well plate with recombinant human TPO antigen detection antibody calibrator and diluent enabling high-throughput accurate quantification of anti-TPO antibodies for diagnosing autoimmune thyroid diseases
Catalog No. 50-000-79780 Supplier Meso Scale Discovery Supplier No. K156VKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP TPO Assay is an electrochemiluminescent immunoassay for measuring thyroid peroxidase autoantibodies in human serum plasma and whole blood It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The assay has a dynamic range of 0 1-1000 U/mL and requires 25 L per well The kit includes a 96-well plate with recombinant human TPO antigen detection antibody calibrator and diluent enabling high-throughput accurate quantification of anti-TPO antibodies for diagnosing autoimmune thyroid diseases