Learn More
Meso Scale Discovery U-PLEX NHP TPO ASY
SDP
Catalog No. 5000079894
Encompass
The Meso Scale Discovery U-PLEX NHP TPO Assay Kit is an electrochemiluminescent immunoassay for measuring thyroid peroxidase autoantibodies in human serum plasma and cell culture supernatant It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from a small sample The kit includes a 96-well plate with TPO antigen detection antibody calibrators controls diluents buffers and seals Sensitivity is 0 2 IU/mL with an upper limit of 1000 IU/mL and a sample volume of 25 L
Catalog No. 50-000-79894 Supplier Meso Scale Discovery Supplier No. K156VKK2
The Meso Scale Discovery U-PLEX NHP TPO Assay Kit is an electrochemiluminescent immunoassay for measuring thyroid peroxidase autoantibodies in human serum plasma and cell culture supernatant It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results from a small sample The kit includes a 96-well plate with TPO antigen detection antibody calibrators controls diluents buffers and seals Sensitivity is 0 2 IU/mL with an upper limit of 1000 IU/mL and a sample volume of 25 L