Learn More
Meso Scale Discovery U-PLEX NHP TPO ASYQP
SDP
Catalog No. 5000080671
Encompass
The U-PLEX NHP TPO Assay (K156VKK) is a specialized immunoassay designed for measuring thrombopoietin (TPO) levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of TPO a key cytokine that regulates platelet production The assay is specifically validated for use with serum plasma and cell culture supernatant samples from non-human primates making it valuable for preclinical research and drug development studies
Catalog No. 50-000-80671 Supplier Meso Scale Discovery Supplier No. K156VKK22
Add to Cart
Add to Cart
The U-PLEX NHP TPO Assay (K156VKK) is a specialized immunoassay designed for measuring thrombopoietin (TPO) levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of TPO a key cytokine that regulates platelet production The assay is specifically validated for use with serum plasma and cell culture supernatant samples from non-human primates making it valuable for preclinical research and drug development studies