Learn More
Meso Scale Discovery U-PLEX NHP TRAIL ASY
SDP
Catalog No. 5000080901
Encompass
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows simultaneous measurement of up to 10 biomarkers related to neurodegenerative diseases and neuroinflammation The assay uses electrochemiluminescence detection technology and is run on Meso Scale Diagnostics QuickPlex SQ 120 instrument The kit includes a 10-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies calibrators and controls Biomarkers measured include A 42 t-tau p-tau -synuclein NFL GFAP sTREM2 YKL-40 MCP-1 and sCD40L The assay requires a low sample volume (25 L) and provides high sensitivity and a broad dynamic range
Catalog No. 50-000-80901 Supplier Meso Scale Discovery Supplier No. K156XTK2
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows simultaneous measurement of up to 10 biomarkers related to neurodegenerative diseases and neuroinflammation The assay uses electrochemiluminescence detection technology and is run on Meso Scale Diagnostics QuickPlex SQ 120 instrument The kit includes a 10-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies calibrators and controls Biomarkers measured include A 42 t-tau p-tau -synuclein NFL GFAP sTREM2 YKL-40 MCP-1 and sCD40L The assay requires a low sample volume (25 L) and provides high sensitivity and a broad dynamic range