Learn More
Meso Scale Discovery U-PLEX NHP TRAIL ASY 25 PL
SDP
Catalog No. 5000080231
Encompass
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) total tau (t-tau) and phosphorylated tau (p-tau181) in human serum and plasma The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators controls 10X diluent read buffer wash buffer and plate seals The assay procedure takes 3 5 hours to complete
Catalog No. 50-000-80231 Supplier Meso Scale Discovery Supplier No. K156XTK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) total tau (t-tau) and phosphorylated tau (p-tau181) in human serum and plasma The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators controls 10X diluent read buffer wash buffer and plate seals The assay procedure takes 3 5 hours to complete