Learn More
Meso Scale Discovery U-PLEX NHP TRAIL ASY384-well
SDP
Catalog No. 5000080273
Encompass
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring neurofilament light chain glial fibrillary acidic protein total tau and phosphorylated tau in human serum and plasma It uses electrochemiluminescence detection for high sensitivity and range The kit includes a 96-well plate ECL-tagged detection antibodies calibrators controls and buffers The assay takes 3 5 hours performs up to 80 tests and lasts 9 months at 2-8 C enabling sensitive biomarker quantification
Catalog No. 50-000-80273 Supplier Meso Scale Discovery Supplier No. K256XTK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP Trail Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring neurofilament light chain glial fibrillary acidic protein total tau and phosphorylated tau in human serum and plasma It uses electrochemiluminescence detection for high sensitivity and range The kit includes a 96-well plate ECL-tagged detection antibodies calibrators controls and buffers The assay takes 3 5 hours performs up to 80 tests and lasts 9 months at 2-8 C enabling sensitive biomarker quantification