Learn More
Meso Scale Discovery U-PLEX NHP TRAIL ASYQP 25 PL
SDP
Catalog No. 5000078630
Encompass
The U-PLEX NHP TRAIL Assay (K156XTK) is a multiplex immunoassay designed for measuring TRAIL (TNF-related apoptosis-inducing ligand) levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for research applications studying apoptosis pathways and immune responses The kit includes pre-coated plates detection reagents and standards for quantitative analysis of TRAIL protein concentrations in serum plasma and cell culture samples
Catalog No. 50-000-78630 Supplier Meso Scale Discovery Supplier No. K156XTK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP TRAIL Assay (K156XTK) is a multiplex immunoassay designed for measuring TRAIL (TNF-related apoptosis-inducing ligand) levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for research applications studying apoptosis pathways and immune responses The kit includes pre-coated plates detection reagents and standards for quantitative analysis of TRAIL protein concentrations in serum plasma and cell culture samples