Learn More
Meso Scale Discovery U-PLEX NHP TRAIL ASYQP
SDP
Catalog No. 5000080902
Encompass
The U-PLEX NHP TRAIL Assay (K156XTK) is a multiplex immunoassay designed for measuring TRAIL (TNF-related apoptosis-inducing ligand) levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for research applications studying apoptosis pathways and immune responses The kit includes pre-coated plates detection reagents and standards for quantitative analysis of TRAIL biomarkers in serum plasma and cell culture samples from non-human primate models
Catalog No. 50-000-80902 Supplier Meso Scale Discovery Supplier No. K156XTK21
The U-PLEX NHP TRAIL Assay (K156XTK) is a multiplex immunoassay designed for measuring TRAIL (TNF-related apoptosis-inducing ligand) levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity and specificity for research applications studying apoptosis pathways and immune responses The kit includes pre-coated plates detection reagents and standards for quantitative analysis of TRAIL biomarkers in serum plasma and cell culture samples from non-human primate models