Learn More
Meso Scale Discovery U-PLEX NHP VEGF-a ASY
SDP
Catalog No. 5000080135
Encompass
The Meso Scale Discovery U-PLEX NHP VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of VEGF-A in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity for VEGF-A with no cross-reactivity to other growth factors It requires 50 L or less sample volume and can be multiplexed with other U-PLEX assays The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and VEGF-A calibrator
Catalog No. 50-000-80135 Supplier Meso Scale Discovery Supplier No. K156UVK2
The Meso Scale Discovery U-PLEX NHP VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of VEGF-A in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity for VEGF-A with no cross-reactivity to other growth factors It requires 50 L or less sample volume and can be multiplexed with other U-PLEX assays The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and VEGF-A calibrator