Learn More
Meso Scale Discovery U-PLEX NHP VEGF-a ASY384-well
SDP
Catalog No. 5000081752
Encompass
The Meso Scale Discovery U-PLEX NHP VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows simultaneous measurement of VEGF-A levels across 384 samples in just 3 5 hours The kit has a lower limit of detection of 0 31 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-81752 Supplier Meso Scale Discovery Supplier No. K256UVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent It allows simultaneous measurement of VEGF-A levels across 384 samples in just 3 5 hours The kit has a lower limit of detection of 0 31 pg/mL and an upper limit of 10 000 pg/mL