Learn More
Meso Scale Discovery U-PLEX NHP YKL-40 ASY 25 PL
SDP
Catalog No. 5000080713
Encompass
The Meso Scale Discovery U-PLEX NHP YKL-40 Assay Kit is a multiplex immunoassay that quantitatively measures YKL-40 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows simultaneous measurement of YKL-40 along with other biomarkers The kit has high performance and reliability for research use
Catalog No. 50-000-80713 Supplier Meso Scale Discovery Supplier No. K156VLK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP YKL-40 Assay Kit is a multiplex immunoassay that quantitatively measures YKL-40 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows simultaneous measurement of YKL-40 along with other biomarkers The kit has high performance and reliability for research use