Learn More
Meso Scale Discovery U-PLEX NHP YKL-40 ASY
SDP
Catalog No. 5000080716
Encompass
The Meso Scale Discovery U-PLEX NHP YKL-40 Assay Kit is a multiplex immunoassay for measuring YKL-40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls It measures YKL-40 and S100A12 simultaneously aiding research in diseases like inflammatory bowel disease liver fibrosis and cancer The assay provides rapid reliable YKL-40 quantification in 3 5 hours
Catalog No. 50-000-80716 Supplier Meso Scale Discovery Supplier No. K156VLK2
The Meso Scale Discovery U-PLEX NHP YKL-40 Assay Kit is a multiplex immunoassay for measuring YKL-40 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and controls It measures YKL-40 and S100A12 simultaneously aiding research in diseases like inflammatory bowel disease liver fibrosis and cancer The assay provides rapid reliable YKL-40 quantification in 3 5 hours