Learn More
Meso Scale Discovery U-PLEX NHP YKL-40 ASY384-well
SDP
Catalog No. 5000081753
Encompass
The Meso Scale Discovery U-PLEX NHP YKL-40 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of YKL-40 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is compatible with MSD MULTI-ARRAY and MULTI-SPOT microplates The kit includes a plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a typical sensitivity of 0 31 ng/mL and an upper limit of quantification of 10 000 ng/mL
Catalog No. 50-000-81753 Supplier Meso Scale Discovery Supplier No. K256VLK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP YKL-40 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of YKL-40 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and is compatible with MSD MULTI-ARRAY and MULTI-SPOT microplates The kit includes a plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a typical sensitivity of 0 31 ng/mL and an upper limit of quantification of 10 000 ng/mL