Learn More
Meso Scale Discovery U-PLEX Obesity Combo 1 ms
SDP
Catalog No. 5000080331
Encompass
The U-PLEX Obesity Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit designed to measure biomarkers related to obesity and metabolic disorders in mouse samples The kit contains reagents to measure leptin insulin PAI-1 total and resistin simultaneously from a single sample It utilizes MSD s electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals This multiplex assay allows efficient analysis of obesity biomarkers in mouse models
Catalog No. 50-000-80331 Supplier Meso Scale Discovery Supplier No. K15300K1
The U-PLEX Obesity Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit designed to measure biomarkers related to obesity and metabolic disorders in mouse samples The kit contains reagents to measure leptin insulin PAI-1 total and resistin simultaneously from a single sample It utilizes MSD s electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals This multiplex assay allows efficient analysis of obesity biomarkers in mouse models