Learn More
Meso Scale Discovery U-PLEX Obesity Combo 1 rt
SDP
Catalog No. 5000082487
Encompass
The Meso Scale Discovery U-PLEX Obesity Combo 1 Rat Kit is a multiplex immunoassay kit for the simultaneous measurement of leptin insulin and C-peptide in rat serum plasma and tissue culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and stop solution The electrochemiluminescent assay allows sensitive detection of the analytes in a small sample volume The kit has a testing time of 3 5 hours and a dynamic range of up to 4 logs It provides a reproducible and robust tool for obesity and metabolic research in rat models
Catalog No. 50-000-82487 Supplier Meso Scale Discovery Supplier No. K15310K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Obesity Combo 1 Rat Kit is a multiplex immunoassay kit for the simultaneous measurement of leptin insulin and C-peptide in rat serum plasma and tissue culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and stop solution The electrochemiluminescent assay allows sensitive detection of the analytes in a small sample volume The kit has a testing time of 3 5 hours and a dynamic range of up to 4 logs It provides a reproducible and robust tool for obesity and metabolic research in rat models